missing translation for 'onlineSavingsMsg'
Learn More

beta-Bromostyrene (cis- and trans- mixture) 95.0+%, TCI America™

Catalog No. b065525g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B065525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B065525G Supplier TCI America Supplier No. B065525G
Only null left

Chemical Identifiers

CAS 103-64-0
Molecular Formula C8H7Br
Molecular Weight (g/mol) 183.048
MDL Number MFCD00000185
InChI Key YMOONIIMQBGTDU-VOTSOKGWSA-N
Synonym 2-bromovinyl benzene, beta-bromostyrene, styryl bromide, 2-bromoethenyl benzene, bromostyrolene, 1-bromo-2-phenylethene, bromostyrol, bromstyrole, hyacinth base, 1-bromo-2-phenylethylene
PubChem CID 5314126
IUPAC Name [(E)-2-bromoethenyl]benzene
SMILES C1=CC=C(C=C1)C=CBr

Specifications

Color Yellow
Quantity 25 g
Formula Weight 183.05
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material beta-Bromostyrene (cis- and trans- mixture)
EINECSNumber (3)-0036
RTECSNumber WL3850000
TSCA Yes
Recommended Storage Refrigerator