missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Alanyl-L-histidine 98.0+%, TCI America™
$36.57 - $105.89
Chemical Identifiers
| CAS | 305-84-0 |
|---|---|
| Molecular Formula | C9H14N4O3 |
| Molecular Weight (g/mol) | 226.236 |
| MDL Number | MFCD00005207 |
| InChI Key | CQOVPNPJLQNMDC-ZETCQYMHSA-N |
| Synonym | l-carnosine, carnosine, beta-alanyl-l-histidine, karnozin, karnozzn, ignotine, l-ignotine, n-2-m, unii-8ho6pvn24w, nalpha-beta-alanyl-l-histidine |
| PubChem CID | 439224 |
| ChEBI | CHEBI:15727 |
| IUPAC Name | (2S)-2-(3-aminopropanoylamino)-3-(1H-imidazol-5-yl)propanoic acid |
| SMILES | C1=C(NC=N1)CC(C(=O)O)NC(=O)CCN |
Chemical Identifiers
| 305-84-0 | |
| 226.236 | |
| CQOVPNPJLQNMDC-ZETCQYMHSA-N | |
| 439224 | |
| (2S)-2-(3-aminopropanoylamino)-3-(1H-imidazol-5-yl)propanoic acid |
| C9H14N4O3 | |
| MFCD00005207 | |
| l-carnosine, carnosine, beta-alanyl-l-histidine, karnozin, karnozzn, ignotine, l-ignotine, n-2-m, unii-8ho6pvn24w, nalpha-beta-alanyl-l-histidine | |
| CHEBI:15727 | |
| C1=C(NC=N1)CC(C(=O)O)NC(=O)CCN |
Specifications
| 305-84-0 | |
| White | |
| MFCD00005207 | |
| l-carnosine, carnosine, beta-alanyl-l-histidine, karnozin, karnozzn, ignotine, l-ignotine, n-2-m, unii-8ho6pvn24w, nalpha-beta-alanyl-l-histidine | |
| C1=C(NC=N1)CC(C(=O)O)NC(=O)CCN | |
| 226.236 | |
| CHEBI:15727 | |
| ≥98.0% (HPLC,T) | |
| beta-Alanyl-L-histidine |
| 260°C | |
| C9H14N4O3 | |
| 1 g | |
| CQOVPNPJLQNMDC-ZETCQYMHSA-N | |
| (2S)-2-(3-aminopropanoylamino)-3-(1H-imidazol-5-yl)propanoic acid | |
| 439224 | |
| 226.24 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-0409&(9)-1557
RTECSNumber : MS3080000
TSCA : Yes