missing translation for 'onlineSavingsMsg'
Learn More

beta-Alanyl-L-histidine 98.0+%, TCI America™

Catalog No. A02221G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A02221G 1 g
A02225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A02221G Supplier TCI America Supplier No. A02221G

Chemical Identifiers

CAS 305-84-0
Molecular Formula C9H14N4O3
Molecular Weight (g/mol) 226.236
MDL Number MFCD00005207
InChI Key CQOVPNPJLQNMDC-ZETCQYMHSA-N
Synonym l-carnosine, carnosine, beta-alanyl-l-histidine, karnozin, karnozzn, ignotine, l-ignotine, n-2-m, unii-8ho6pvn24w, nalpha-beta-alanyl-l-histidine
PubChem CID 439224
ChEBI CHEBI:15727
IUPAC Name (2S)-2-(3-aminopropanoylamino)-3-(1H-imidazol-5-yl)propanoic acid
SMILES C1=C(NC=N1)CC(C(=O)O)NC(=O)CCN

Specifications

Melting Point 260°C
Color White
Quantity 1 g
Formula Weight 226.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material beta-Alanyl-L-histidine
EINECSNumber (5)-0409&(9)-1557
RTECSNumber MS3080000
TSCA Yes