missing translation for 'onlineSavingsMsg'
Learn More

beta-Alanyl-L-histidine, 95%

CAS: 305-84-0 | C9H14N4O3 | 226.236 g/mol

$108.21 - $436.03

Chemical Identifiers

CAS 305-84-0
Molecular Formula C9H14N4O3
Molecular Weight (g/mol) 226.236
MDL Number MFCD00005207
InChI Key CQOVPNPJLQNMDC-ZETCQYMHSA-N
Synonym l-carnosine, carnosine, beta-alanyl-l-histidine, karnozin, karnozzn, ignotine, l-ignotine, n-2-m, unii-8ho6pvn24w, nalpha-beta-alanyl-l-histidine
PubChem CID 439224
ChEBI CHEBI:15727
IUPAC Name (2S)-2-(3-aminopropanoylamino)-3-(1H-imidazol-5-yl)propanoic acid
SMILES C1=C(NC=N1)CC(C(=O)O)NC(=O)CCN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6566806
View Documents
Thermo Scientific Chemicals
H6566806
5 g
Each for $108.21
Only null left
 
AAH6566814
View Documents
Thermo Scientific Chemicals
H6566814
25 g
Each for $436.03
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

305-84-0
226.236
CQOVPNPJLQNMDC-ZETCQYMHSA-N
439224
(2S)-2-(3-aminopropanoylamino)-3-(1H-imidazol-5-yl)propanoic acid
C9H14N4O3
MFCD00005207
l-carnosine, carnosine, beta-alanyl-l-histidine, karnozin, karnozzn, ignotine, l-ignotine, n-2-m, unii-8ho6pvn24w, nalpha-beta-alanyl-l-histidine
CHEBI:15727
C1=C(NC=N1)CC(C(=O)O)NC(=O)CCN

Specifications

253°C
White
MFCD00005207
l-carnosine, carnosine, beta-alanyl-l-histidine, karnozin, karnozzn, ignotine, l-ignotine, n-2-m, unii-8ho6pvn24w, nalpha-beta-alanyl-l-histidine
C1=C(NC=N1)CC(C(=O)O)NC(=O)CCN
5 g
439224
226.24
Crystalline
305-84-0
C9H14N4O3
87671
CQOVPNPJLQNMDC-ZETCQYMHSA-N
(2S)-2-(3-aminopropanoylamino)-3-(1H-imidazol-5-yl)propanoic acid
226.236
CHEBI:15727
95%
beta-Alanyl-L-histidine
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 206-169-9

RTECSNumber : MS3080000

TSCA : Yes

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

RUO – Research Use Only