missing translation for 'onlineSavingsMsg'
Learn More

beta-Alanine tert-Butyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. p-7118063
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27725G 5 g
A277225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27725G Supplier TCI America Supplier No. A27725G
Only null left

Chemical Identifiers

CAS 58620-93-2
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00058258
InChI Key DOMTZTVJNZKUNX-UHFFFAOYSA-N
Synonym tert-butyl 3-aminopropanoate hydrochloride, h-beta-ala-otbu.hcl, beta-alanine tert-butyl ester hydrochloride, h-beta-ala-otbu hcl, beta-alanine t-butyl ester hydrochloride, 3-amino-propionic acid tert-butyl ester hydrochloride, h-?-ala-otbu.hcl, h-, a-ala-otbu.hcl, h-beta-ala-otbu hydrochloride, tert-butyl 3-aminopropanoate-hcl
PubChem CID 16218890
IUPAC Name tert-butyl 3-aminopropanoate hydrochloride
SMILES Cl.CC(C)(C)OC(=O)CCN

Specifications

Melting Point 169°C
Color White-Yellow
Quantity 5 g
Formula Weight 181.66
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material beta-Alanine tert-Butyl Ester Hydrochloride
TSCA No
Recommended Storage Freezer