missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Alanine tert-Butyl Ester Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 169°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 181.66 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | beta-Alanine tert-Butyl Ester Hydrochloride |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |