missing translation for 'onlineSavingsMsg'
Learn More

beta-Alanine tert-butyl ester hydrochloride, 95%

CAS: 58620-93-2 | C7H16ClNO2 | 181.66 g/mol

$127.44 - $1324.86

Chemical Identifiers

CAS 58620-93-2
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00058258
InChI Key DOMTZTVJNZKUNX-UHFFFAOYSA-N
Synonym tert-butyl 3-aminopropanoate hydrochloride, tert-butyl 3-aminopropanoate-hcl, beta-alanine tert-butyl ester hydrochloride, beta-alanine tert-butyl ester hydrochloride, beta-alanine t-butyl ester hydrochloride, beta-alanine t-butyl ester hydrochloride, h-?-ala-otbu.hcl, h-?-ala-otbu.hcl, a-ala-otbu.hcl, h-beta-ala-otbu hydrochloride, h-beta-ala-otbu hydrochloride
PubChem CID 16218890
IUPAC Name tert-butyl 3-aminopropanoate;hydrochloride
SMILES Cl.CC(C)(C)OC(=O)CCN
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6434606
View Documents
Thermo Scientific Chemicals
H6434606
5 g
Each for $127.44
Only null left
 
AAH6434614
View Documents
Thermo Scientific Chemicals
H6434614
25 g
Each for $449.16
Only null left
 
AAH6434622
View Documents
Thermo Scientific Chemicals
H6434622
100 g
Each for $1,324.86
Only null left
 
Description

Description

Useful intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful intermediate for pharmaceutical and organic synthesis.

Solubility
Clear solution (0.3g in 2ml MeOH).

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

58620-93-2
181.66
DOMTZTVJNZKUNX-UHFFFAOYSA-N
16218890
Cl.CC(C)(C)OC(=O)CCN
C7H16ClNO2
MFCD00058258
tert-butyl 3-aminopropanoate hydrochloride, tert-butyl 3-aminopropanoate-hcl, beta-alanine tert-butyl ester hydrochloride, beta-alanine tert-butyl ester hydrochloride, beta-alanine t-butyl ester hydrochloride, beta-alanine t-butyl ester hydrochloride, h-?-ala-otbu.hcl, h-?-ala-otbu.hcl, a-ala-otbu.hcl, h-beta-ala-otbu hydrochloride, h-beta-ala-otbu hydrochloride
tert-butyl 3-aminopropanoate;hydrochloride

Specifications

167°C to 169°C
C7H16ClNO2
MFCD00058258
tert-butyl 3-aminopropanoate hydrochloride, tert-butyl 3-aminopropanoate-hcl, beta-alanine tert-butyl ester hydrochloride, beta-alanine tert-butyl ester hydrochloride, beta-alanine t-butyl ester hydrochloride, beta-alanine t-butyl ester hydrochloride, h-?-ala-otbu.hcl, h-?-ala-otbu.hcl, a-ala-otbu.hcl, h-beta-ala-otbu hydrochloride, h-beta-ala-otbu hydrochloride
DOMTZTVJNZKUNX-UHFFFAOYSA-N
tert-butyl 3-aminopropanoate;hydrochloride
181.66
181.66
Solid
58620-93-2
H2NCH2CH2COOC(CH3)3·HCl
6660122
Clear solution (0.3g in 2ml MeOH).
Cl.CC(C)(C)OC(=O)CCN
5 g
16218890
95%
beta-Alanine tert-butyl ester hydrochloride
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

RUO – Research Use Only