missing translation for 'onlineSavingsMsg'
Learn More

beta-Alanine Methyl Ester Hydrochloride 98.0+%, TCI America™

Supplier:  TCI America A260325G

Quantity 25 g

 View more versions of this product

Catalog No. A260325G


Only null left

Description

Description

Specifications

Chemical Identifiers

3196-73-4
139.58
XPGRZDJXVKFLHQ-UHFFFAOYSA-N
2734767
Cl.COC(=O)CCN
C4H10ClNO2
MFCD00039060
methyl 3-aminopropionate hydrochloride, methyl 3-aminopropanoate hydrochloride, beta-alanine methyl ester hydrochloride, beta-alanine methyl ester hcl, h-beta-ala-ome.hcl, h-, a-ala-ome.hcl, h-beta-ala-ome hydrochloride, b-alanine methyl ester hydrochloride, 3-aminopropanoic acid methyl ester hydrochloride, 3-aminopropionic acid methyl ester hydrochloride
methyl 3-aminopropanoate hydrochloride

Specifications

108°C
3196-73-4
C4H10ClNO2
methyl 3-aminopropionate hydrochloride, methyl 3-aminopropanoate hydrochloride, beta-alanine methyl ester hydrochloride, beta-alanine methyl ester hcl, h-beta-ala-ome.hcl, h-, a-ala-ome.hcl, h-beta-ala-ome hydrochloride, b-alanine methyl ester hydrochloride, 3-aminopropanoic acid methyl ester hydrochloride, 3-aminopropionic acid methyl ester hydrochloride
Cl.COC(=O)CCN
139.58
2734767
≥98.0% (T)
beta-Alanine Methyl Ester Hydrochloride
White-Yellow
MFCD00039060
XPGRZDJXVKFLHQ-UHFFFAOYSA-N
methyl 3-aminopropanoate hydrochloride
25 g
139.58
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions