missing translation for 'onlineSavingsMsg'
Learn More

beta-Alanine methyl ester hydrochloride, 98%

Catalog No. AAH2748314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2748314 25 g
AAH2748306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2748314 Supplier Thermo Scientific Chemicals Supplier No. H2748314

CAS: 3196-73-4 | C4H10ClNO2 | 139.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3196-73-4
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.58
MDL Number MFCD00039060
InChI Key XPGRZDJXVKFLHQ-UHFFFAOYSA-N
Synonym methyl 3-aminopropionate hydrochloride, methyl 3-aminopropanoate hydrochloride, beta-alanine methyl ester hydrochloride, beta-alanine methyl ester hcl, h-beta-ala-ome.hcl, h-, a-ala-ome.hcl, h-beta-ala-ome hydrochloride, b-alanine methyl ester hydrochloride, 3-aminopropanoic acid methyl ester hydrochloride, 3-aminopropionic acid methyl ester hydrochloride
PubChem CID 2734767
IUPAC Name methyl 3-aminopropanoate;hydrochloride
SMILES Cl.COC(=O)CCN

Specifications

Melting Point 102°C to 104°C
Linear Formula H2NCH2CH2CO2CH3·HCl
Beilstein 3556748
Quantity 25 g
Formula Weight 139.58
Percent Purity 98%
Sensitivity Hygroscopic
Chemical Name or Material beta-Alanine methyl ester hydrochloride
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only