missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Alanine Ethyl Ester Hydrochloride 98.0+%, TCI America™
$55.84 - $123.21
Chemical Identifiers
| CAS | 4244-84-2 |
|---|---|
| Molecular Formula | C5H12ClNO2 |
| Molecular Weight (g/mol) | 153.61 |
| MDL Number | MFCD00012909 |
| InChI Key | RJCGNNHKSNIUAT-UHFFFAOYSA-N |
| Synonym | ethyl 3-aminopropanoate hydrochloride, beta-alanine ethyl ester hydrochloride, ethyl beta-alaninate hydrochloride, h-beta-ala-oet.hcl, h-b-ala-oet hcl, h-beta-ala-oet hcl, h-?-ala-oet.hcl, b-alanine ethyl ester hydrochloride, ethyl 3-aminopropionate hydrochloride, beta-alanine ethyl ester hcl |
| PubChem CID | 458475 |
| IUPAC Name | ethyl 3-aminopropanoate hydrochloride |
| SMILES | Cl.CCOC(=O)CCN |
Chemical Identifiers
| 4244-84-2 | |
| 153.61 | |
| RJCGNNHKSNIUAT-UHFFFAOYSA-N | |
| 458475 | |
| Cl.CCOC(=O)CCN |
| C5H12ClNO2 | |
| MFCD00012909 | |
| ethyl 3-aminopropanoate hydrochloride, beta-alanine ethyl ester hydrochloride, ethyl beta-alaninate hydrochloride, h-beta-ala-oet.hcl, h-b-ala-oet hcl, h-beta-ala-oet hcl, h-?-ala-oet.hcl, b-alanine ethyl ester hydrochloride, ethyl 3-aminopropionate hydrochloride, beta-alanine ethyl ester hcl | |
| ethyl 3-aminopropanoate hydrochloride |
Specifications
| 69°C | |
| White | |
| MFCD00012909 | |
| RJCGNNHKSNIUAT-UHFFFAOYSA-N | |
| ethyl 3-aminopropanoate hydrochloride | |
| 153.61 | |
| 153.61 | |
| Crystalline Powder |
| 4244-84-2 | |
| C5H12ClNO2 | |
| ethyl 3-aminopropanoate hydrochloride, beta-alanine ethyl ester hydrochloride, ethyl beta-alaninate hydrochloride, h-beta-ala-oet.hcl, h-b-ala-oet hcl, h-beta-ala-oet hcl, h-?-ala-oet.hcl, b-alanine ethyl ester hydrochloride, ethyl 3-aminopropionate hydrochloride, beta-alanine ethyl ester hcl | |
| Cl.CCOC(=O)CCN | |
| 10 g | |
| 458475 | |
| ≥98.0% (T) | |
| beta-Alanine Ethyl Ester Hydrochloride |
Safety and Handling
TSCA : No