missing translation for 'onlineSavingsMsg'
Learn More

beta-Alanine Ethyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. A131010G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A131010G 10 g
A131025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A131010G Supplier TCI America Supplier No. A131010G
Only null left

Chemical Identifiers

CAS 4244-84-2
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00012909
InChI Key RJCGNNHKSNIUAT-UHFFFAOYSA-N
Synonym ethyl 3-aminopropanoate hydrochloride, beta-alanine ethyl ester hydrochloride, ethyl beta-alaninate hydrochloride, h-beta-ala-oet.hcl, h-b-ala-oet hcl, h-beta-ala-oet hcl, h-?-ala-oet.hcl, b-alanine ethyl ester hydrochloride, ethyl 3-aminopropionate hydrochloride, beta-alanine ethyl ester hcl
PubChem CID 458475
IUPAC Name ethyl 3-aminopropanoate hydrochloride
SMILES Cl.CCOC(=O)CCN

Specifications

Melting Point 69°C
Color White
Quantity 10 g
Formula Weight 153.61
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material beta-Alanine Ethyl Ester Hydrochloride