missing translation for 'onlineSavingsMsg'
Learn More

beta-Alanine ethyl ester hydrochloride, 98%

Catalog No. AAA1727709
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1727709 10 g
AAA1727718 50 g
AAA1727730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1727709 Supplier Thermo Scientific Chemicals Supplier No. A1727709
Only null left

CAS: 4244-84-2 | C5H12ClNO2 | 153.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4244-84-2
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00012909
InChI Key RJCGNNHKSNIUAT-UHFFFAOYSA-N
Synonym ethyl 3-aminopropanoate hydrochloride, beta-alanine ethyl ester hydrochloride, ethyl beta-alaninate hydrochloride, h-beta-ala-oet.hcl, h-b-ala-oet hcl, h-beta-ala-oet hcl, h-?-ala-oet.hcl, b-alanine ethyl ester hydrochloride, ethyl 3-aminopropionate hydrochloride, beta-alanine ethyl ester hcl
PubChem CID 458475
IUPAC Name ethyl 3-aminopropanoate;hydrochloride
SMILES Cl.CCOC(=O)CCN

Specifications

Melting Point 67°C to 72°C
Beilstein 3559095
Quantity 10 g
Formula Weight 153.61
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material beta-Alanine ethyl ester hydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-203-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only