missing translation for 'onlineSavingsMsg'
Learn More

beta-Alanine benzyl ester p-toluenesulfonate, 98%

Catalog No. p-7024280
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1865009 10 g
AAA1865018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1865009 Supplier Thermo Scientific Chemicals Supplier No. A1865009

CAS: 27019-47-2 | C10H14NO2 | 180.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27019-47-2
Molecular Formula C10H14NO2
Molecular Weight (g/mol) 180.23
MDL Number MFCD00039061
InChI Key CANCPUBPPUIWPX-UHFFFAOYSA-O
Synonym beta-alanine benzyl ester p-toluenesulfonate, beta-alanine benzyl ester p-toluenesulfonate salt, h-beta-ala-obzl.tosoh, h-beta-ala-obzl tos, benzyl 3-aminopropanoate 4-methylbenzenesulfonate, betaala-obzl.tosoh, pubchem12070, acmc-209gum, h-, a-ala-obzl.tosoh, h-ala-obzl p-tosylate
PubChem CID 13502100
SMILES [NH3+]CCC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 138°C to 139°C
IUPAC Name benzyl 3-aminopropanoate;4-methylbenzenesulfonic acid
Quantity 10 g
Formula Weight 351.42
Percent Purity 98%
Chemical Name or Material beta-Alanine benzyl ester p-toluenesulfonate
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only