missing translation for 'onlineSavingsMsg'
Learn More

beta-Alaninamide hydrochloride, 96%

CAS: 64017-81-8 | C3H9ClN2O | 124.568 g/mol

$104.67 - $398.96

Chemical Identifiers

CAS 64017-81-8
Molecular Formula C3H9ClN2O
Molecular Weight (g/mol) 124.568
MDL Number MFCD00060747
InChI Key GAGJMOQGABUOBK-UHFFFAOYSA-N
PubChem CID 22222288
IUPAC Name 3-aminopropanamide;hydrochloride
SMILES C(CN)C(=O)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6536506
View Documents
Thermo Scientific Chemicals
H6536506
5 g
Each for $104.67
Only null left
 
AAH6536514
View Documents
Thermo Scientific Chemicals
H6536514
25 g
Each for $398.96
Only null left
 
Description

Description

beta-Alaninamide hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
beta-Alaninamide hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol.

Notes
Air Sensitive. Store away from air. Protect from heat. Store under dry inert gas. Incompatible with heat , air and oxidizing agents.
Specifications

Chemical Identifiers

64017-81-8
124.568
GAGJMOQGABUOBK-UHFFFAOYSA-N
3-aminopropanamide;hydrochloride
C3H9ClN2O
MFCD00060747
22222288
C(CN)C(=O)N.Cl

Specifications

149°C to 154°C
C3H9ClN2O
3909917
Soluble in methanol.
C(CN)C(=O)N.Cl
5 g
22222288
Air sensitive
Solid
64017-81-8
MFCD00060747
3-aminopropanamide hydrochloride, 3-aminopropionamide hydrochloride, 3-aminopropanamide hcl, beta-alaninamide hydrochloride, h-beta-ala-nh2.hcl, h-beta-ala-nh2 hcl, 3-amino-propionamide hydrochloride, h-ala-nh2hcl, propanamide, 3-amino-, monohydrochloride, h-beta-ala-nh2hcl
GAGJMOQGABUOBK-UHFFFAOYSA-N
3-aminopropanamide;hydrochloride
124.568
124.57
96%
beta-Alaninamide hydrochloride
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

RUO – Research Use Only