missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals beta-Adrenergic Receptor Kinase1 Inhibitor

Catalog No. AAJ64615LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64615LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64615LB0 Supplier Thermo Scientific Chemicals Supplier No. J64615LB0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

A selective inhibitor of -ARK1 (--Adrenergic Receptor Kinase 1)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24269-96-3
Molecular Formula C12H9NO6
Synonym β-ARK1 Inhibitor; GRK2 Inhibitor

Specifications

Quantity 5 mg
Description Yellow
Content And Storage -30°C to -10°C
For Use With (Application) A selective inhibitor of →ARK1 (→-Adrenergic Receptor Kinase 1)
Formula Weight 263.20
Sensitivity Light Sensitive
Product Type beta-Adrenergic Receptor Kinase1 Inhibitor
Form Powder

RUO – Research Use Only