missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ beta-1 Adrenergic R/ADRB1 Peptide
Specifications
Specifications
| Gene ID (Entrez) | 153 |
| Species | Human |
| Product Line | Novus |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB600-978 |
| Gene Alias | ADRB1RRHR, adrenergic, beta-1-, receptor, B1AR, beta-1 adrenergic receptor, Beta-1 adrenoceptor, Beta-1 adrenoreceptor, BETA1AR |
| Gene Symbol | ADRB1 |
| Quantity | 0.1 mg |
| Research Category | GPCR |