missing translation for 'onlineSavingsMsg'
Learn More

CMD (Cotton Mouton Diagnostics) αBET Consumable Pack

Catalog No. 12333032
Change view
Click to view available options
Format:
Array Cartridge, Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
12333032 Array Cartridge, Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12333032 Supplier CMD (Cotton Mouton Diagnostics) Supplier No. CMDA0006C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The CMD αBET consumable pack contains 25 four-channel cartridges plus 5 vials of CMD nanorods, which when coupled with FUJIFILM Wako's PYROSTAR ES-F single test kit, provides users with a total of 100 tests.

  • 25 heat-sterilized cartridges with four testing channels
  • Endotoxin-free
  • Proprietary CMD nanorods
  • No special storage required

Specifications

Format Array Cartridge, Vial