Learn More
SHOWA™ Nitri Master™ Gloves
Catalog No. 189992933
Change view
Click to view available options
:
Size 7
Catalog No.
189992933 Size 7
1 options
Catalog No. 189992933 Supplier SHOWA™ Supplier No. NM1107
Only null left
Ergonomic design reduces fatigue and enhances comfort
Combine nitrile stength, durability and chemical resistance in a flexible, low-modulus glove that is lower in cost than traditional gloves. Ideal for asbestos abatement, chemical processing, construction, hospital cleanup, maintenance, metal machining and oil use, paint mixing, pesticide handling and manufacturing, meat and poultry processing, vegetable and animal fat handling, laboratory and agricultural applications.
  • Tractor-tread finish gives a vise-like grip in wet and dry situations
  • Channels away hazardous chemicals from the glove surface
  • Unsupported glove has a two-layer design with a built-in safety feature: two-tone green color provides a wear indicator signaling when to change to a new pair
  • Unlined; Length, 13 in. (33cm); Thickness, 11mil
Compliance
Comply with 21CFR170–1991.
1 Glove is made using only listed materials and levels outlined in the Code of Federal Regulations (21CFR), parts 170–199, and is acceptable for use in both food and pharmaceutical applications.
Specifications
Size 7
Length (English) 12 in.
Length (Metric) 30.5 cm