missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ 4000P Nitrile Coated Cotton Gloves

Catalog No. p-141382
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
1139526B Medium
1139526C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1139526B Supplier SHOWA™ Supplier No. 4000P09
Only null left

A versatile alternative to light cotton or leather gloves. Nitrile coating on the palm/fingers is resistant to oil and grease for enhanced grip capabilities.

  • Nitrile coated for maximum wear, durability, and flexibility
  • Enhanced grip in dry or oily conditions
  • Resistant to punctures, abrasion, oil, and grease
  • Two-piece interlock knit lining for comfort
  • Knit wrist for easy donning

Specifications

Coated Area Palm, Fingers, Knuckles
Coating Material Nitrile
Color Gray, Natural
Exterior Finish No Texture
Hand Specific
Material Cotton
Product Type Dip-coated Gloves
Size Medium
For Use With (Application) Assembly, Inspection, Shipping/Receiving, Light Fabrication, and Material Handling
Glove Cuff Color Red
Glove Cuff Style Knit
Packaging Type Polybag
Series 4000P
Length (English) 10.2 in.
Length (Metric) 26 cm
Show More Show Less