missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHOWA™ 6797 Chemical Resistant Neoprene Gloves, Extended Sleeve
Description
- Extended-arm protection
- Protection against acids, caustics, oils, greases and many solvents
- Protection against physical hazards such as abrasion and cut
- Cotton liner is cool and comfortable in hot conditions and keeps wearer warm in cold conditions
- Ideal applications include:
- Chemical spray and treatment
- Coating preparation
- Laboratory, pharma and analysis
- Painting and spray workshops
Specifications
Specifications
| Certifications/Compliance | 21 CFR 170-199 |
| Certifications/Compliance (Europe) | Cat III CE 0321, EN 407:3242XX |
| Applications | Laboratory |
| Color | Black |
| Latex Free | Yes |
| Material | Neoprene |
| Product Type | Neoprene Gloves |
| Size | 10 |
| EN388 Standard | EN 388:3111X |
| EN ISO 374-1 | Type A |
| Show More |