missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHOWA™ 6784 Chemical Resistant Neoprene Gloves, Cotton Liner
Description
- Chemical resistant against acids, caustics, oils, greases and many solvents
- Protection against physical hazards such as abrasion and cut
- Rough grip for wet applications
- Cotton liner keeps cool and comfortable in hot conditions or warm in cold conditions
- Ideal for biotechnology, cytostatic, life sciences, pharmaceuticals and API
Specifications
Specifications
| Certifications/Compliance | 21 CFR 170-199 |
| Certifications/Compliance (Europe) | Cat III CE 0321, EN 407:4342XX |
| Color | Black |
| Latex Free | Yes |
| Material | Neoprene |
| Size | 10 |
| EN388 Standard | EN 388:3111X |
| EN 421 | No |
| EN ISO 374-1 | Type A |
| EN ISO 374-1 CHEMICAL LETTER CODES | AJKLMT |
| Show More |