missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ 6780 Chemical Resistant Neoprene Gloves, Cotton Liner

Catalog No. 2870413
Change view
Click to view available options
:
12 in. (30 cm) Long; Smooth
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
2870413 12 in. (30 cm) Long; Smooth
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2870413 Supplier SHOWA™ Supplier No. 678010
Only null left

Smooth surface chemical resistant neoprene glove with soft cotton liner protects against acids, chemicals and caustics.

  • For use in hot and cold environments
  • Chemical resistant against acids, chemicals and caustics
  • Rough grip ensures precision in wet or greasy environments
  • Soft cotton liner to maintain hand temperature and absorb perspiration
  • Rugged neoprene coating
  • Ideal for applications involving chemicals, engineering, metallurgy, oil/gas and utilities

Specifications

Certifications/Compliance 21 CFR 170-199
Certifications/Compliance (Europe) Cat III CE 2777, EN 407:X2XXXX, EN 420:2003+A1:2009
Color Black
Latex Free Yes
Material Neoprene
Size 10
EN388 Standard EN 388:2111XP
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AKLMPST
Glove Cuff Style Gauntlet
Grip Design Smooth
Liner Material Cotton
Lining Lined
Model 6780-10
Series 6780
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less