missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ Heavy Duty PVC Coated Seamless Knit Cotton Gloves

Catalog No. 1139516A
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
1139516A Small
1139516B Medium
1139516C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 1139516A Supplier SHOWA™ Supplier No. 960S08

PVC-coating minimizes the absorption of water, oil, and grease with a rough-texture to enhance grip capabilities.

  • PVC-coating ensures breathability
  • Easy donning and doffing with slip-on cuff
  • Minimizes absorption of water, oil, and grease
  • Textured to enhance grip capabilities

Specifications

Coated Area Full Glove
Construction Seamless Knit
Coating Material PVC
Color Yellow
Exterior Finish Fully Textured
Hand Specific
Product Type Seamless Knit Gloves
Size Small
For Use With (Application) Chemical Spray/Treatment, Utility Work, Intricate Parts Handling, and Oil-Coated Pieces
Glove Cuff Style Continuous Knit
Liner Material Cotton
Packaging Type Polybag
Series 960
Show More Show Less