Learn More
SHOWA™ Viton™ and Viton II Gloves
Supplier:  SHOWA™ 890XL10
Catalog No. 2870543
Only null left
Description
Description
Provide highest chemical-resistance barrier to aromatic hydrocarbons such as benzene, toluene, or xylene.
  • Unsupported, unlined gloves made of Viton over butyl are heavy-duty and flexible
  • Smooth finish and rolled cuffs
  • Thicker Viton II gloves also provde a barrier to most chlorinated solvents and aliphatic hydrocarbons
Product Suggestions
Product Suggestions