missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ PVC Triple-Dipped Oil-Resistant ESD Gloves

Catalog No. 19019897
Change view
Click to view available options
Size:
10
9
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19019897 10
19019896 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19019897 Supplier SHOWA™ Supplier No. 660ESDXL10

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gloves are designed for handling flammable or explosive materials, featuring an anti-static liner and coating for protection in extreme environments.

  • Triple-dipped PVC coating protects against chemicals, while also remaining flexible
  • Anti-static liner and coating prevents sparks caused by static electricity, helping to avoid accidental explosions
  • Impermeable design is ideal for working in damp or greasy environments
  • Ergonomic shape provides extra comfort to reduce hand fatigue
  • Special hydrocarbon treatment
  • Actibacterial and anti-odor treated
  • Seamless knit liner
  • Scalloped edge
  • Rough finish

Specifications

Coating Material PVC
Color Black
Exterior Finish Fully Textured
Hand Specific
Product Type ESD Gloves
Size 10
For Use With (Application) Handling Flammable, or Explosive Materials
Glove Cuff Style Gauntlet
Liner Material Cotton/Polyester Blend
Series 660ESD
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less