missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals BES sodium salt, 99%

Catalog No. ac439030250
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
Catalog No. Quantity
AC439030250 25 g
1 options

Catalog No. AC439030250

Supplier: Thermo Scientific Chemicals 439030250

Only null left

CAS: 66992-27-6 | C6H14NO5S·Na | 235.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 66992-27-6
Molecular Formula C6H14NO5S·Na
Molecular Weight (g/mol) 235.24
InChI Key CFQLQLSIZOWFNV-UHFFFAOYSA-M
Synonym bes sodium salt, sodium 2-bis 2-hydroxyethyl amino ethanesulfonate, n,n-bis 2-hydroxyethyl-2-aminoethanesulfonic acid sodium salt, sodium 2-bis 2-hydroxyethyl amino ethane-1-sulfonate, ethanesulfonic acid, 2-bis 2-hydroxyethyl amino-, monosodium salt, 2-bis 2-hydroxyethyl amino ethanesulfonic acid, sodium salt, bes na, bes sodium salt buffer, potassium c6h14no5s
PubChem CID 23671640
IUPAC Name sodium;2-[bis(2-hydroxyethyl)amino]ethanesulfonate
SMILES C(CO)N(CCO)CCS(=O)(=O)[O-].[Na+]

Specifications

Melting Point 155.6°C to 156.5°C
Color White
CAS Min % 98.5
CAS Max % 100.0
Physical Form Crystalline Powder or Powder
Quantity 25 g
Assay Percent Range 99%
Formula Weight 235.24
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass Bottle
Chemical Name or Material BES sodium salt
Show More Show Less
Hazard Statement GHS H Statement:
Precautionary Statement GHS P Statement:
Do not get in eyes, on skin, or on clothing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only