Learn More
Berkshire™ CapSure-VP™ Wipers
Engineered for superior surface cleaning ability
Supplier:  Berkshire™ CPSVP.0909.8
 View more versions of this product
Catalog No. 17-897-214
Only null left
Explore available promotions
Description
Description
  • Continuous filament polyester knit with sealed edges
  • Retains 93% of particles initially captured
  • Minimal particles, fibers, ions and extractable
  • Low airborne partical release
  • Cleanroom laundered
Specifications
Specifications
Cleanroom Wipe
22.86 cm
22.86 cm
IPA, Acetone and other solvents
131
Sealed
For use in ISO Class 3 and higher cleanroom environments, applying and removing cleaning and disinfecting solutions
150 Sheets/Pk., 8 Pk./Cs.
9 in.
9 in.
Polyester Knit
Autoclavable
RoHS 2002/05, REACH
9 x 9 in. (22.86 x 22.86 cm)
For cleaning of medical device products
Non-sterile
Product Suggestions
Product Suggestions
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'