missing translation for 'onlineSavingsMsg'
Learn More

Bergenin 98.0+%, TCI America™

Catalog No. B43491G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43491G 1 g
B43495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43491G Supplier TCI America Supplier No. B43491G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 477-90-7
Molecular Formula C14H16O9
Molecular Weight (g/mol) 328.273
MDL Number MFCD00133120
InChI Key YWJXCIXBAKGUKZ-HJJNZUOJSA-N
PubChem CID 66065
ChEBI CHEBI:69499
IUPAC Name (2R,3S,4S,4aR,10bS)-3,4,8,10-tetrahydroxy-2-(hydroxymethyl)-9-methoxy-3,4,4a,10b-tetrahydro-2H-pyrano[3,2-c]isochromen-6-one
SMILES COC1=C(C=C2C(=C1O)C3C(C(C(C(O3)CO)O)O)OC2=O)O

Specifications

Melting Point 240°C
Color White
Quantity 1 g
Physical Form Crystalline Powder
Formula Weight 328.27
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material Bergenin
RTECSNumber UQ0337200
TSCA No
Recommended Storage Refrigerator