missing translation for 'onlineSavingsMsg'
Learn More

Bergapten 98.0+%, TCI America™

Catalog No. B28401G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28401G 1 g
B28405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28401G Supplier TCI America Supplier No. B28401G

Chemical Identifiers

CAS 484-20-8
Molecular Formula C12H8O4
Molecular Weight (g/mol) 216.192
InChI Key BGEBZHIAGXMEMV-UHFFFAOYSA-N
Synonym bergapten, 5-methoxypsoralen, bergaptene, heraclin, majudin, 4-methoxy-7h-furo 3,2-g chromen-7-one, bergaptan, psoraderm, 5-mop, o-methylbergaptol
PubChem CID 2355
ChEBI CHEBI:18293
IUPAC Name 4-methoxyfuro[3,2-g]chromen-7-one
SMILES COC1=C2C=CC(=O)OC2=CC3=C1C=CO3

Specifications

Melting Point 191°C
Color White-Yellow
Quantity 1 g
Formula Weight 216.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bergapten
RTECSNumber LV1300000
TSCA No