missing translation for 'onlineSavingsMsg'
Learn More

Berberine Chloride Hydrate 98.0+%, TCI America™

Catalog No. B04505G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B04505G 5 g
B045025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B04505G Supplier TCI America Supplier No. B04505G

Chemical Identifiers

CAS 141433-60-5
Molecular Formula C20H18ClNO4
Molecular Weight (g/mol) 371.82
MDL Number MFCD00011939
InChI Key VKJGBAJNNALVAV-UHFFFAOYSA-M
Synonym berberine chloride hydrate, 9,10-dimethoxy-5,6-dihydro-1,3 dioxolo 4,5-g isoquinolino 3,2-a isoquinolin-7-ium chloride hydrate, c20h18no4.cl.h2o, benzo g-1,3-benzodioxolo 5,6-a quinolizinium, 5,6-dihydro-9,10-dimethoxy-, chloride, monohydrate, kyoberin tn, 9,10-dimethoxy-5,6-dihydro-2h-1,3-dioxolano 4,5-g isoquinolino 3,2-a isoquinol ine, chloride, hydrate, berberinechloridehydrate, berberine chloride xhydrate, berberine chloride hydrate jp17
PubChem CID 155074
IUPAC Name 16,17-dimethoxy-5,7-dioxa-13λ⁵-azapentacyclo[11.8.0.0²,¹⁰.0⁴,⁸.0¹⁵,²⁰]henicosa-1(13),2,4(8),9,14,16,18,20-octaen-13-ylium chloride
SMILES [Cl-].COC1=C(OC)C2=C[N+]3=C(C=C2C=C1)C1=CC2=C(OCO2)C=C1CC3

Specifications

Melting Point 145°C
Quantity 5 g
Formula Weight 371.82
Percent Purity ≥98.0% (HPLC,T)
Physical Form Powder
Chemical Name or Material Berberine Chloride Hydrate
RTECSNumber DR9866400
TSCA Yes