missing translation for 'onlineSavingsMsg'
Learn More

Berberine chloride hydrate, 96%, water <17%

Catalog No. p-7058279
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
AAL0380706 5 g
AAL0380714 25 g
2 options

Catalog No. AAL0380706

Supplier: Thermo Scientific Chemicals L0380706

Only null left

CAS: 141433-60-5 | C20H18ClNO4 | 371.82 g/mol

Primarily for the treatment of gastroenteritis, dysentery and other intestinal infections and conjunctivitis, purulent otitis media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Primarily for the treatment of gastroenteritis, dysentery and other intestinal infections and conjunctivitis, purulent otitis media.

Solubility
Soluble in water.

Notes
Light sensitive. Store in the dark. Store away from oxidizing agents, light.

Chemical Identifiers

CAS 141433-60-5
Molecular Formula C20H18ClNO4
Molecular Weight (g/mol) 371.82
MDL Number MFCD00011939
InChI Key VKJGBAJNNALVAV-UHFFFAOYSA-M
Synonym berberine chloride hydrate, 9,10-dimethoxy-5,6-dihydro-1,3 dioxolo 4,5-g isoquinolino 3,2-a isoquinolin-7-ium chloride hydrate, c20h18no4.cl.h2o, benzo g-1,3-benzodioxolo 5,6-a quinolizinium, 5,6-dihydro-9,10-dimethoxy-, chloride, monohydrate, kyoberin tn, 9,10-dimethoxy-5,6-dihydro-2h-1,3-dioxolano 4,5-g isoquinolino 3,2-a isoquinol ine, chloride, hydrate, berberinechloridehydrate, berberine chloride xhydrate, berberine chloride hydrate jp17
PubChem CID 155074
IUPAC Name 16,17-dimethoxy-5,7-dioxa-13λ⁵-azapentacyclo[11.8.0.0²,¹⁰.0⁴,⁸.0¹⁵,²⁰]henicosa-1(13),2,4(8),9,14,16,18,20-octaen-13-ylium chloride
SMILES [Cl-].COC1=C(OC)C2=C[N+]3=C(C=C2C=C1)C1=CC2=C(OCO2)C=C1CC3

Specifications

Melting Point ∽200°C (decomposition)
Quantity 5 g
Beilstein 3836585
Sensitivity Light Sensitive
Merck Index 14,1154
Solubility Information Soluble in water.
Formula Weight 371.82 (Anhydrous)
Percent Purity 97%
Assay water <17%
Chemical Name or Material Berberine chloride hydrate
EINECSNumber 211-195-9
RTECSNumber DR9866400
TSCA Yes

RUO – Research Use Only