missing translation for 'onlineSavingsMsg'
Learn More

Berberine chloride hydrate, 96%, water <17%

Catalog No. AAL0380714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0380714 25 g
AAL0380706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0380714 Supplier Thermo Scientific Chemicals Supplier No. L0380714

CAS: 141433-60-5 | C20H18ClNO4 | 371.82 g/mol

Primarily for the treatment of gastroenteritis, dysentery and other intestinal infections and conjunctivitis, purulent otitis media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Primarily for the treatment of gastroenteritis, dysentery and other intestinal infections and conjunctivitis, purulent otitis media.

Solubility
Soluble in water.

Notes
Light sensitive. Store in the dark. Store away from oxidizing agents, light.

Chemical Identifiers

CAS 141433-60-5
Molecular Formula C20H18ClNO4
Molecular Weight (g/mol) 371.82
MDL Number MFCD00011939
InChI Key VKJGBAJNNALVAV-UHFFFAOYSA-M
Synonym berberine chloride hydrate, 9,10-dimethoxy-5,6-dihydro-1,3 dioxolo 4,5-g isoquinolino 3,2-a isoquinolin-7-ium chloride hydrate, c20h18no4.cl.h2o, benzo g-1,3-benzodioxolo 5,6-a quinolizinium, 5,6-dihydro-9,10-dimethoxy-, chloride, monohydrate, kyoberin tn, 9,10-dimethoxy-5,6-dihydro-2h-1,3-dioxolano 4,5-g isoquinolino 3,2-a isoquinol ine, chloride, hydrate, berberinechloridehydrate, berberine chloride xhydrate, berberine chloride hydrate jp17
PubChem CID 155074
IUPAC Name 16,17-dimethoxy-5,7-dioxa-13λ⁵-azapentacyclo[11.8.0.0²,¹⁰.0⁴,⁸.0¹⁵,²⁰]henicosa-1(13),2,4(8),9,14,16,18,20-octaen-13-ylium chloride
SMILES [Cl-].COC1=C(OC)C2=C[N+]3=C(C=C2C=C1)C1=CC2=C(OCO2)C=C1CC3

Specifications

Melting Point ∽200°C (decomposition)
Quantity 25 g
Beilstein 3836585
Sensitivity Light Sensitive
Merck Index 14,1154
Solubility Information Soluble in water.
Formula Weight 371.82 (Anhydrous)
Percent Purity 97%
Assay water <17%
Chemical Name or Material Berberine chloride hydrate
EINECSNumber 211-195-9
RTECSNumber DR9866400
TSCA Yes

RUO – Research Use Only