missing translation for 'onlineSavingsMsg'
Learn More

Benzyltrimethylammonium dichloroiodate, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 268600050

 View more versions of this product

Catalog No. AC268600050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Benzyltrimethylammonium dichloroiodate
114971-52-7
100.0
99%
C10H16Cl2IN
MFCD00075259
15,25; 08,434; 13,266; 15,278; 16,289; 17,298; 05,571
Solubility in water: soluble in water. Other solubilities: soluble in methylene chloride
C[N+](C)(C)CC1=CC=CC=C1.Cl[I-]Cl
2724680
99%
99%
98.5
Authentic
Glass bottle
C6H5CH2N+(CH3)3Cl2I-
5g
benzyltrimethylammonium dichloroiodate, benzyltrimethylammonium dichloroiodide, btma-icl2, benzyl trimethylammonium dichloroiodate, benzyltrimethylazanium; dichloroiodanuide, pubchem21920, benzyltrimethylammoniumdichloroiodate
PPDJNZTUDFPAHX-UHFFFAOYSA-N
348.04
348.04

Chemical Identifiers

114971-52-7
348.04
PPDJNZTUDFPAHX-UHFFFAOYSA-N
2724680
C10H16Cl2IN
MFCD00075259
benzyltrimethylammonium dichloroiodate, benzyltrimethylammonium dichloroiodide, btma-icl2, benzyl trimethylammonium dichloroiodate, benzyltrimethylazanium; dichloroiodanuide, pubchem21920, benzyltrimethylammoniumdichloroiodate
C[N+](C)(C)CC1=CC=CC=C1.Cl[I-]Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only