missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyltriethylammonium Iodide 98.0+%, TCI America™
$83.89 - $229.29
Chemical Identifiers
| CAS | 5400-94-2 |
|---|---|
| Molecular Formula | C13H22IN |
| Molecular Weight (g/mol) | 319.23 |
| MDL Number | MFCD00043178 |
| InChI Key | JWLJBISFJGEYMT-UHFFFAOYSA-M |
| PubChem CID | 122756 |
| IUPAC Name | benzyltriethylazanium iodide |
| SMILES | [I-].CC[N+](CC)(CC)CC1=CC=CC=C1 |
Chemical Identifiers
| 5400-94-2 | |
| 319.23 | |
| JWLJBISFJGEYMT-UHFFFAOYSA-M | |
| benzyltriethylazanium iodide |
| C13H22IN | |
| MFCD00043178 | |
| 122756 | |
| [I-].CC[N+](CC)(CC)CC1=CC=CC=C1 |
Specifications
| 5400-94-2 | |
| White | |
| MFCD00043178 | |
| JWLJBISFJGEYMT-UHFFFAOYSA-M | |
| benzyltriethylazanium iodide | |
| 122756 | |
| ≥98.0% (T) | |
| Benzyltriethylammonium Iodide |
| 171°C | |
| C13H22IN | |
| 5 g | |
| [I-].CC[N+](CC)(CC)CC1=CC=CC=C1 | |
| 319.23 | |
| 319.23 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-2694
TSCA : No