missing translation for 'onlineSavingsMsg'
Learn More

Benzylsulfonyl Fluoride 98.0+%, TCI America™

Catalog No. p-7118828
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10625G 5 g
B106225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B10625G Supplier TCI America Supplier No. B10625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 329-98-6
Molecular Formula C7H7FO2S
Molecular Weight (g/mol) 174.189
MDL Number MFCD00007424
InChI Key YBYRMVIVWMBXKQ-UHFFFAOYSA-N
Synonym phenylmethylsulfonyl fluoride, pmsf, benzenemethanesulfonyl fluoride, benzylsulfonyl fluoride, alpha-toluenesulfonyl fluoride, benzylsulphonyl fluoride, alpha-toluenesulphonyl fluoride, phenylmethylsulfonylfluoride, phenylmethylsulphonyl fluoride, .alpha.-toluenesulfonyl fluoride
PubChem CID 4784
ChEBI CHEBI:8102
IUPAC Name phenylmethanesulfonyl fluoride
SMILES C1=CC=C(C=C1)CS(=O)(=O)F

Specifications

Melting Point 94°C
Color White-Yellow
Boiling Point 85°C
Quantity 5 g
UN Number 2923
Formula Weight 174.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzylsulfonyl Fluoride
RTECSNumber XT8040000
TSCA Yes