missing translation for 'onlineSavingsMsg'
Learn More

Benzylsulfonyl Fluoride 98.0+%, TCI America™

Catalog No. p-7122968
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B34735G 5 g
B347325G 25 g
2 options

Catalog No. B34735G

Supplier: TCI America B34735G

Only null left

Chemical Identifiers

CAS 329-98-6
Molecular Formula C7H7FO2S
Molecular Weight (g/mol) 174.189
MDL Number MFCD00007424
InChI Key YBYRMVIVWMBXKQ-UHFFFAOYSA-N
Synonym phenylmethylsulfonyl fluoride, pmsf, benzenemethanesulfonyl fluoride, benzylsulfonyl fluoride, alpha-toluenesulfonyl fluoride, benzylsulphonyl fluoride, alpha-toluenesulphonyl fluoride, phenylmethylsulfonylfluoride, phenylmethylsulphonyl fluoride, .alpha.-toluenesulfonyl fluoride
PubChem CID 4784
ChEBI CHEBI:8102
IUPAC Name phenylmethanesulfonyl fluoride
SMILES C1=CC=C(C=C1)CS(=O)(=O)F

Specifications

Melting Point 94°C
Color White-Yellow
Boiling Point 85°C
Quantity 5 g
UN Number 2923
Formula Weight 174.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzylsulfonyl Fluoride [for Biochemical Research]
RTECSNumber XT8040000
TSCA Yes