missing translation for 'onlineSavingsMsg'
Learn More

Benzylphosphonic acid, 97%

CAS: 6881-57-8 | C7H7O3P | 170.11 g/mol

$60.38 - $780.26

Chemical Identifiers

CAS 6881-57-8
Molecular Formula C7H7O3P
Molecular Weight (g/mol) 170.11
MDL Number MFCD00039519
InChI Key OGBVRMYSNSKIEF-UHFFFAOYSA-L
Synonym benzyl-phosphonic acid, phosphonic acid, phenylmethyl, phenylmethylphosphonic acid, phosphonic acid, p-phenylmethyl, phenyl-methanephosphonic acid anion, acmc-1b2tf, phenylmethanephosphonic acid, alpha-toluenephosphonic acid, benzylphosphonic acid, bmk1-h2
PubChem CID 81312
IUPAC Name benzylphosphonic acid
SMILES [O-]P([O-])(=O)CC1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1385003
View Documents
Thermo Scientific Chemicals
A1385003
1 g
Each for $60.38
Only null left
 
AAA1385006
View Documents
Thermo Scientific Chemicals
A1385006
5 g
Each for $195.95
Only null left
 
AAA1385014
View Documents
Thermo Scientific Chemicals
A1385014
25 g
Each for $780.26
Only null left
 
Description

Description

Structural mimic of phosphotyrosine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6881-57-8
170.11
OGBVRMYSNSKIEF-UHFFFAOYSA-L
81312
[O-]P([O-])(=O)CC1=CC=CC=C1
C7H7O3P
MFCD00039519
benzyl-phosphonic acid, phosphonic acid, phenylmethyl, phenylmethylphosphonic acid, phosphonic acid, p-phenylmethyl, phenyl-methanephosphonic acid anion, acmc-1b2tf, phenylmethanephosphonic acid, alpha-toluenephosphonic acid, benzylphosphonic acid, bmk1-h2
benzylphosphonic acid

Specifications

6881-57-8
C7H7O3P
1 g
2519318
OGBVRMYSNSKIEF-UHFFFAOYSA-L
benzylphosphonic acid
81312
97%
166°C to 172°C
MFCD00039519
UN3261
benzyl-phosphonic acid, phosphonic acid, phenylmethyl, phenylmethylphosphonic acid, phosphonic acid, p-phenylmethyl, phenyl-methanephosphonic acid anion, acmc-1b2tf, phenylmethanephosphonic acid, alpha-toluenephosphonic acid, benzylphosphonic acid, bmk1-h2
[O-]P([O-])(=O)CC1=CC=CC=C1
170.11
172.11
Benzylphosphonic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only