missing translation for 'onlineSavingsMsg'
Learn More

Benzylphosphonic acid, 97%

Catalog No. AAA1385014
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1385014 25 g
AAA1385003 1 g
AAA1385006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1385014 Supplier Thermo Scientific Chemicals Supplier No. A1385014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6881-57-8 | C7H7O3P | 170.11 g/mol

Structural mimic of phosphotyrosine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6881-57-8
Molecular Formula C7H7O3P
Molecular Weight (g/mol) 170.11
MDL Number MFCD00039519
InChI Key OGBVRMYSNSKIEF-UHFFFAOYSA-L
Synonym benzyl-phosphonic acid, phosphonic acid, phenylmethyl, phenylmethylphosphonic acid, phosphonic acid, p-phenylmethyl, phenyl-methanephosphonic acid anion, acmc-1b2tf, phenylmethanephosphonic acid, alpha-toluenephosphonic acid, benzylphosphonic acid, bmk1-h2
PubChem CID 81312
IUPAC Name benzylphosphonic acid
SMILES [O-]P([O-])(=O)CC1=CC=CC=C1

Specifications

Melting Point 166°C to 172°C
Quantity 25 g
UN Number UN3261
Beilstein 2519318
Formula Weight 172.11
Percent Purity 97%
Chemical Name or Material Benzylphosphonic acid
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only