missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+/-)-Benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester, 97%
Description
(±)-Z-α-Phosphonoglycine trimethyl ester is the starting material for the synthesis of (Z)-dehydroamino acids by Wittig-Horner reaction with aldehydes, the dehydroamino acids can be stereo selectively reduced to amino acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(±)-Z-α-Phosphonoglycine trimethyl ester is the starting material for the synthesis of (Z)-dehydroamino acids by Wittig-Horner reaction with aldehydes, the dehydroamino acids can be stereo selectively reduced to amino acids.
Solubility
Soluble in methanol.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
(±)-Z-α-Phosphonoglycine trimethyl ester is the starting material for the synthesis of (Z)-dehydroamino acids by Wittig-Horner reaction with aldehydes, the dehydroamino acids can be stereo selectively reduced to amino acids.
Solubility
Soluble in methanol.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 76°C to 80°C |
| Quantity | 5 g |
| Solubility Information | Soluble in methanol. |
| Formula Weight | 331.27 |
| Percent Purity | 97% |
| Chemical Name or Material | (+/-)-Benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only