missing translation for 'onlineSavingsMsg'
Learn More

Benzyldimethylstearylammonium Chloride Hydrate 98.0+%, TCI America™

$98.79 - $264.70

Chemical Identifiers

CAS 122-19-0
Molecular Formula C27H50ClN
Molecular Weight (g/mol) 424.154
MDL Number MFCD00011776
InChI Key SFVFIFLLYFPGHH-UHFFFAOYSA-M
Synonym Octadecyldimethylbenzylammonium Chloride, Benzyldimethyloctadecylammonium Chloride
PubChem CID 31204
IUPAC Name benzyl-dimethyl-octadecylazanium;chloride
SMILES CCCCCCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1.[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B129725G
View Documents
TCI America
B129725G
25 g
Each for $98.79
Only null left
 
B1297100G
View Documents
TCI America
B1297100G
100 g
Each for $264.70
Only null left
 
Description

Description

Specifications

Chemical Identifiers

122-19-0
424.154
SFVFIFLLYFPGHH-UHFFFAOYSA-M
31204
CCCCCCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1.[Cl-]
C27H50ClN
MFCD00011776
Octadecyldimethylbenzylammonium Chloride, Benzyldimethyloctadecylammonium Chloride
benzyl-dimethyl-octadecylazanium;chloride

Specifications

122-19-0
C27H50ClN
25 g
SFVFIFLLYFPGHH-UHFFFAOYSA-M
benzyl-dimethyl-octadecylazanium;chloride
31204
≥98.0% (T)
Benzyldimethylstearylammonium Chloride Hydrate
White
MFCD00011776
Octadecyldimethylbenzylammonium Chloride, Benzyldimethyloctadecylammonium Chloride
CCCCCCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1.[Cl-]
424.154
424.15
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0326&(3)-2694

RTECSNumber : BO7000000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'