missing translation for 'onlineSavingsMsg'
Learn More

Benzyldimethylstearylammonium Chloride Hydrate 98.0+%, TCI America™

Catalog No. B1297100G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1297100G 100 g
B129725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1297100G Supplier TCI America Supplier No. B1297100G

Chemical Identifiers

CAS 122-19-0
Molecular Formula C27H50ClN
Molecular Weight (g/mol) 424.154
MDL Number MFCD00011776
InChI Key SFVFIFLLYFPGHH-UHFFFAOYSA-M
Synonym Octadecyldimethylbenzylammonium Chloride, Benzyldimethyloctadecylammonium Chloride
PubChem CID 31204
IUPAC Name benzyl-dimethyl-octadecylazanium;chloride
SMILES CCCCCCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1.[Cl-]

Specifications

Color White
Quantity 100 g
Formula Weight 424.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyldimethylstearylammonium Chloride Hydrate
EINECSNumber (3)-0326&(3)-2694
RTECSNumber BO7000000
TSCA Yes