Learn More
Benzylboronic acid pinacol ester, 96%
CAS: 87100-28-5 | C13H19BO2 | 218.103 g/mol
Supplier: Thermo Scientific Chemicals H3252306
Description
Benzylboronic acid pinacol ester finds application in Pd (0)-mediated [11 C] carbonylation reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzylboronic acid pinacol ester finds application in Pd (0)-mediated [11 C] carbonylation reaction.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
| Benzylboronic acid pinacol ester | |
| 0.98 | |
| C13H19BO2 | |
| 5 g | |
| Insoluble in water. | |
| B1(OC(C(O1)(C)C)(C)C)CC2=CC=CC=C2 | |
| 218.103 | |
| 218.1 |
| 87100-28-5 | |
| 65°C (0.15 mmHg) | |
| MFCD05663841 | |
| benzylboronic acid pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-phenylmethyl, acmc-209qek, benzylboronic pinacol ester, benzyl boronic acid pinacol ester, benzylboronic acid, pinacol ester, 2-benzyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 4,4,5,5-tetramethyl-2-phenylmethyl-1,3,2-dioxaborolane | |
| YCNQPAVKQPLZRS-UHFFFAOYSA-N | |
| 2-benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 3864964 | |
| 96% |
Chemical Identifiers
| 87100-28-5 | |
| 218.103 | |
| YCNQPAVKQPLZRS-UHFFFAOYSA-N | |
| 3864964 | |
| B1(OC(C(O1)(C)C)(C)C)CC2=CC=CC=C2 |
| C13H19BO2 | |
| MFCD05663841 | |
| benzylboronic acid pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-phenylmethyl, acmc-209qek, benzylboronic pinacol ester, benzyl boronic acid pinacol ester, benzylboronic acid, pinacol ester, 2-benzyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 4,4,5,5-tetramethyl-2-phenylmethyl-1,3,2-dioxaborolane | |
| 2-benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Safety and Handling
TSCA : No
RUO – Research Use Only