missing translation for 'onlineSavingsMsg'
Learn More

Benzylacetone, 99%

Catalog No. p-65954
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105830050 5 mL Glass bottle
AC105832500 250 mL Glass bottle
AC105830010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC105830050 Supplier Thermo Scientific Chemicals Supplier No. 105830050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2550-26-7 | C10H12O | 148.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2550-26-7
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.2
MDL Number MFCD00008790
InChI Key AKGGYBADQZYZPD-UHFFFAOYSA-N
Synonym benzylacetone, 4-phenyl-2-butanone, 2-butanone, 4-phenyl, 4-penylbutan-2-one, 1-phenyl-3-butanone, methyl phenethyl ketone, benzyl acetone, methyl 2-phenylethyl ketone, phenethyl methyl ketone, methyl phenylethyl ketone
PubChem CID 17355
IUPAC Name 4-phenylbutan-2-one
SMILES CC(=O)CCC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9875g/mL
Boiling Point 235°C to 237°C
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5100 to 1.513
Linear Formula C6H5CH2CH2COCH3
Quantity 5 mL
Beilstein 07, 314
Specific Gravity 0.9875
Solubility Information Solubility in water: practically insoluble in water.. Other solubilities: soluble in ethanol,diethyl ether,chloroform,,very soluble in acetone
Formula Weight 148.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Benzylacetone, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 219-847-4
RTECSNumber EL9600000
TSCA TSCA

RUO – Research Use Only