missing translation for 'onlineSavingsMsg'
Learn More

Benzyl viologen dichloride, 97%

Catalog No. AAH6683606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6683606 5 g
AAH6683603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6683606 Supplier Thermo Scientific Chemicals Supplier No. H6683606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1102-19-8 | C24H22Cl2N2 | 409.354 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1102-19-8
Molecular Formula C24H22Cl2N2
Molecular Weight (g/mol) 409.354
MDL Number MFCD00011991
InChI Key NLOIIDFMYPFJKP-UHFFFAOYSA-L
Synonym 1,1'-dibenzyl-4,4'-bipyridinium dichloride, benzyl viologen, benzylviologen chloride, benzyl viologen dichloride, n,n'-dibenzyl-gamma,gamma'-dipyridylium dichloride, 1,1'-bis phenylmethyl-4,4'-bipyridinium dichloride, 4,4'-bipyridinium, 1,1'-dibenzyl-, dichloride, 4,4'-bipyridinium, 1,1'-bis phenylmethyl-, dichloride, 1,1'-dibenzyl-4,4'-bipyridine-1,1'-diium chloride, 1-benzyl-4-1-benzylpyridin-1-ium-4-yl pyridin-1-ium dichloride
PubChem CID 14195
IUPAC Name 1-benzyl-4-(1-benzylpyridin-1-ium-4-yl)pyridin-1-ium;dichloride
SMILES C1=CC=C(C=C1)C[N+]2=CC=C(C=C2)C3=CC=[N+](C=C3)CC4=CC=CC=C4.[Cl-].[Cl-]

Specifications

Melting Point 260°C (decomposition)
Color White
Quantity 5 g
Beilstein 3811585
Formula Weight 409.35
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Benzyl viologen dichloride
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 214-158-5
RTECSNumber DW1930000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only