missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl (S)-(-)-Lactate 98.0+%, TCI America™
Supplier: TCI America L016125G
Specifications
| Benzyl (S)-(-)-Lactate | |
| 17°C | |
| 141°C | |
| MFCD00209546 | |
| benzyl s---lactate, benzyl l-lactate, benzyl s-lactate, s---lactic acid benzyl ester, benzyl 2s-2-hydroxypropanoate, benzyl-l-lactate, s-benzyllactate, s-benzyl lactate, s-lactic acid benzyl ester, 2s-lactic acid benzyl ester | |
| CC(C(=O)OCC1=CC=CC=C1)O | |
| 180.203 | |
| 180.20 | |
| Liquid |
| 56777-24-3 | |
| Yellow | |
| C10H12O3 | |
| 25 g | |
| ZYTLPUIDJRKAAM-QMMMGPOBSA-N | |
| benzyl (2S)-2-hydroxypropanoate | |
| 11052238 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 56777-24-3 | |
| 180.203 | |
| ZYTLPUIDJRKAAM-QMMMGPOBSA-N | |
| 11052238 | |
| CC(C(=O)OCC1=CC=CC=C1)O |
| C10H12O3 | |
| MFCD00209546 | |
| benzyl s---lactate, benzyl l-lactate, benzyl s-lactate, s---lactic acid benzyl ester, benzyl 2s-2-hydroxypropanoate, benzyl-l-lactate, s-benzyllactate, s-benzyl lactate, s-lactic acid benzyl ester, 2s-lactic acid benzyl ester | |
| benzyl (2S)-2-hydroxypropanoate |
Safety and Handling
TSCA : No