missing translation for 'onlineSavingsMsg'
Learn More

Benzyl (S)-(-)-Lactate 98.0+%, TCI America™

Catalog No. L01615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01615G 5 g
L016125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L01615G Supplier TCI America Supplier No. L01615G

Chemical Identifiers

CAS 56777-24-3
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00209546
InChI Key ZYTLPUIDJRKAAM-QMMMGPOBSA-N
Synonym benzyl s---lactate, benzyl l-lactate, benzyl s-lactate, s---lactic acid benzyl ester, benzyl 2s-2-hydroxypropanoate, benzyl-l-lactate, s-benzyllactate, s-benzyl lactate, s-lactic acid benzyl ester, 2s-lactic acid benzyl ester
PubChem CID 11052238
IUPAC Name benzyl (2S)-2-hydroxypropanoate
SMILES CC(C(=O)OCC1=CC=CC=C1)O

Specifications

Melting Point 17°C
Color Yellow
Boiling Point 141°C
Quantity 5 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl (S)-(-)-Lactate