missing translation for 'onlineSavingsMsg'
Learn More

Benzyl (R)-(-)-Glycidyl Ether 98.0+%, TCI America™

Catalog No. B22385G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22385G 5 g
B22381G 1 g
B223825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B22385G Supplier TCI America Supplier No. B22385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14618-80-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00068409
InChI Key QNYBOILAKBSWFG-JTQLQIEISA-N
Synonym r-benzyloxymethyl-oxirane, r---benzyl glycidyl ether, r-2-benzyloxy methyl oxirane, benzyl r---glycidyl ether, r-o-benzylglycidol, r-benzyl glycidyl ether, r---2-benzyloxymethyl oxirane, ccris 6385, --benzyl-r-glycidylether, r---benzyloxymethyloxirane
PubChem CID 159746
IUPAC Name (2R)-2-(phenylmethoxymethyl)oxirane
SMILES C1C(O1)COCC2=CC=CC=C2

Specifications

Color Colorless
Boiling Point 130°C
Quantity 5 g
Formula Weight 164.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl (R)-(-)-Glycidyl Ether
RTECSNumber TX2860030
TSCA No