missing translation for 'onlineSavingsMsg'
Learn More

Benzyl (R)-(-)-glycidyl ether, 98+%

Catalog No. AAL1398006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1398006 5 g
AAL1398003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1398006 Supplier Thermo Scientific Chemicals Supplier No. L1398006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14618-80-5 | C10H12O2 | 164.204 g/mol

Benzyl (R)-(-)-glycidyl ether is used in the preparation of the lactone fragment of compactin and mevinolin. Chiron in the preparation of syn-1,3-polyols, dideoxynucleosides, and a spiroacetal cyanohydrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl (R)-(-)-glycidyl ether is used in the preparation of the lactone fragment of compactin and mevinolin. Chiron in the preparation of syn-1,3-polyols, dideoxynucleosides, and a spiroacetal cyanohydrin.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Chemical Identifiers

CAS 14618-80-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00068409
InChI Key QNYBOILAKBSWFG-JTQLQIEISA-N
Synonym r-benzyloxymethyl-oxirane, r---benzyl glycidyl ether, r-2-benzyloxy methyl oxirane, benzyl r---glycidyl ether, r-o-benzylglycidol, r-benzyl glycidyl ether, r---2-benzyloxymethyl oxirane, ccris 6385, --benzyl-r-glycidylether, r---benzyloxymethyloxirane
PubChem CID 159746
IUPAC Name (2R)-2-(phenylmethoxymethyl)oxirane
SMILES C1C(O1)COCC2=CC=CC=C2

Specifications

Density 1.072
Boiling Point 130°C (0.1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.517
Quantity 5 g
Beilstein 3588399
Solubility Information Soluble in water.
Optical Rotation −5.1° (c=5 in Toluene)
Formula Weight 164.2
Percent Purity ≥98%
Chemical Name or Material Benzyl (R)-(-)-glycidyl ether
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H341-H315-H319
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber TX2860030
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only