missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Phenyl Ketone 98.0+%, TCI America™

Supplier:  TCI America B0435250G

 View more versions of this product

Catalog No. B0435250G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Benzyl Phenyl Ketone
56°C
322°C
MFCD00003081
2-phenylacetophenone, deoxybenzoin, benzyl phenyl ketone, ethanone, 1,2-diphenyl, desoxybenzoin, 1,2-diphenylethan-1-one, phenyl benzyl ketone, benzoin, deoxy, acetophenone, 2-phenyl, deoxy benzoin
O=C(CC1=CC=CC=C1)C1=CC=CC=C1
196.25
196.25
Crystalline Powder
451-40-1
White-Yellow
C14H12O
250 g
OTKCEEWUXHVZQI-UHFFFAOYSA-N
1,2-diphenylethan-1-one
9948
≥98.0% (GC)

Chemical Identifiers

451-40-1
196.25
OTKCEEWUXHVZQI-UHFFFAOYSA-N
9948
O=C(CC1=CC=CC=C1)C1=CC=CC=C1
C14H12O
MFCD00003081
2-phenylacetophenone, deoxybenzoin, benzyl phenyl ketone, ethanone, 1,2-diphenyl, desoxybenzoin, 1,2-diphenylethan-1-one, phenyl benzyl ketone, benzoin, deoxy, acetophenone, 2-phenyl, deoxy benzoin
1,2-diphenylethan-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-1893

RTECSNumber : AM9662500

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions