missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Nicotinate 98.0+%, TCI America™
Specifications
Specifications
| CAS | 94-44-0 |
| Boiling Point | 189°C |
| Molecular Formula | C13H11NO2 |
| Quantity | 25 g |
| MDL Number | MFCD00023584 |
| Synonym | benzyl nicotinate, nicotinic acid benzyl ester, rubriment, pycaril, pykaryl, niacin benzyl ester, 3-pyridinecarboxylic acid, phenylmethyl ester, benzylis nicotinas, nicotinic acid, benzyl ester, nicotinsaeurebenzylester |
| InChI Key | KVYGGMBOZFWZBQ-UHFFFAOYSA-N |
| SMILES | C1=CC=C(C=C1)COC(=O)C2=CN=CC=C2 |
| IUPAC Name | benzyl pyridine-3-carboxylate |
| Molecular Weight (g/mol) | 213.236 |
| Show More |
Safety and Handling
| EINECSNumber | (9)-0957 |
| RTECSNumber | QT0850000 |
| TSCA | Yes |